missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21523-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an endo/exonuclease with 5′-3′ exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5′ end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.Specifications
| EXOG | |
| Polyclonal | |
| Unconjugated | |
| EXOG | |
| Endo G like 1, ENDOGL1, ENDOGL2, Endonuclease G like 1, ENGL, ENGL a, ENGL B, EXOG, Nuclease EXOG, mitochondrial | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 208194, 301062, 9941 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C163, Q9Y2C4 | |
| EXOG | |
| EXOG Fusion Protein Ag16122 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction