missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Exosome component 8 Overexpression Lysate

Product Code. 18233862 Shop All Bio Techne Products
Change view
Click to view available options
:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18233862 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18233862 Supplier Novus Biologicals™ Supplier No. NBL110389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Exosome component 8 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Especificaciones

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC24768 and based on accession number NM_18153. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens exosome component 8 (EXOSC8), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Research Category DNA replication Transcription Translation and Splicing
Protein State Native

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.