missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Exosome Standards (B16F10 cell line)

Product Code. 18692650 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18692650 6 x 100 μg Vials Pack of 6
18684351 2 x 30 μg Vials Pack of 2
18650621 6 x 30 μg Vials Pack of 6
18682591 2 x 100 μg Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18692650 Supplier R&D Systems™ Supplier No. NBP249866600ug
  • ONLINE EXCLUSIVE PRICE
    1565.36 €
    1358.70 € / Pack of 6
    Save 206.66 €
    13% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Product Line Novus
Quantity 6 x 100 μg Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.