missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Exosome Standards (COLO1 cell line)

Product Code. 18694040 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18694040 2 x 30 μg Vials Pack of 2
18655380 6 x 30 μg Vials Pack of 6
18644351 2 x 100 μg Vials Pack of 2
18652591 6 x 100 μg Vials Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18694040 Supplier R&D Systems™ Supplier No. NBP24984560ug
  • ONLINE EXCLUSIVE PRICE
    474.72 €
    416.85 € / Pack of 2
    Save 57.87 €
    12% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Assay Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Product Line Novus
Quantity 2 x 30 μg Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.