Learn More
Memmert™ Extended Overtemperature Protection for SF160
Product Code. 17568115 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF160 Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17568115 SF160 Sterilizer Each
1 options
Product Code. 17568115 Supplier Memmert™ Supplier No. SF160A6
  • ONLINE EXCLUSIVE PRICE
    194.00 €
    192.00 € / Pack of 1
    Save 2.00 €
    1% Off
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
For Use With (Equipment) SF160 Sterilizer
Includes Integrated Pt100 Sensor