Learn More
Memmert™ Extension of DAkkS Factory Calibration for SF55
Product Code. 17588315 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
17588315 Each
1 options
Product Code. 17588315 Supplier Memmert™ Supplier No. SF55E39697
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Sterilizers.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) SF55 Sterilizer
Product Type Factory Calibration Extension