missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Extension of DAkkS Factory Calibration for UN450plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN450plus Oven
Unit Size:
Each
Description
- Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
| For Use With (Equipment) | UN450plus Oven |
| Product Type | Factory Calibration Extension |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction