missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EYA1 Recombinant Rabbit Monoclonal Antibody (24H60L7)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 703582

Product Code. 17166298

  • 481.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with Bovine.

This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EYA1
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P97767, Q99502
EYA1
Peptides corresponding to human EYA1 [1) aa11-aa29 2) aa290-aa303].
100 μg
Primary
Human, Mouse
Antibody
IgG
Western Blot
24H60L7
Unconjugated
EYA1
BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770
Rabbit
Protein A
RUO
14048, 2138
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.