missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EYA2 Recombinant Rabbit Monoclonal Antibody (23GB5350), Invitrogen™
Description
Functions both as protein phosphatase and as transcriptional coactivator for SIX1, and probably also for SIX2, SIX4 and SIX5. Tyrosine phosphatase that dephosphorylates 'Tyr- 142' of histone H2AX (H2AXY142ph) and promotes efficient DNA repair via the recruitment of DNA repair complexes containing MDC1. 'Tyr-142' phosphorylation of histone H2AX plays a central role in DNA repair and acts as a mark that distinguishes between apoptotic and repair responses to genotoxic stress. Its function as histone phosphatase may contribute to its function in transcription regulation during organogenesis. Plays an important role in hypaxial muscle development together with SIX1 and DACH2; in this it is functionally redundant with EYA1.
Specifications
Specifications
| Antigen | EYA2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB5350 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | EYA2 |
| Gene Accession No. | O00167, O08575 |
| Gene Alias | Drosophila-type eyes absent 2-like protein; Eab1; EYA transcriptional coactivator and phosphatase 2; Eya2; eyes absent 2; eyes absent 2 homolog; eyes absent 2 homolog (Drosophila); eyes absent 2 isoform a; eyes absent homolog 2 |
| Show More |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?