missing translation for 'onlineSavingsMsg'
Learn More

Sartorius F-12 (HAM) Nutrient Mixture

Product Code. 31908414 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908414 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908414 Supplier Sartorius Supplier No. 010951A
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells.

  • F-12 has been used for the serum-free growth of CHO cell cultures.
  • F-12 also supports the serum-supplemented growth of other mammalian cells, including chondrocytes, rat hepatocytes, and rat prostate epithelial cells.
  • Modified as follows:
    • With L-Glutamine
    • With Glucose
    • With Phenol Red
    • With Sodium Pyruvate
    • With Sodium Bicarbonate

Specifications

Form Liquid
Product Type Ham's F-12 Cell Culture Media
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.