missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antigen Rat, APC-Cy7, Clone: BM8.1, enQuireBio™
Rat Monoclonal Antibody
288.75 € - 619.50 €
Specifications
| Antigen | F4/80 Antigen |
|---|---|
| Clone | BM8.1 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15992348
|
EnquireBio
QAB68-APC7-25ug |
25 μg |
288.75 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15982348
|
EnquireBio
QAB68-APC7-100ug |
100 μg |
619.50 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| F4/80 Antigen | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| 13733 | |
| IgG2b κ | |
| Protein A/G Purified |
| BM8.1 | |
| Flow Cytometry | |
| APC-Cyanine7 | |
| RUO | |
| XP_006523664 | |
| Adgre1 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title