missing translation for 'onlineSavingsMsg'
Learn More

FAAH-IN-2, MedChemExpress

Product Code. 16470688
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470688 5 g 5g
16411999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16470688 Supplier Medchem Express Supplier No. HY79511
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FAAH-IN-2 (O-Desmorpholinopropyl Gefitinib) is a potent FAAH(fatty acid amide hydrolase) inhibitor extracted from Patent WO/2008/100977A2.

FAAH-IN-2 (O-Desmorpholinopropyl Gefitinib) is a potent FAAH(fatty acid amide hydrolase) inhibitor extracted from Patent WO/2008/100977A2.

Specifications

Chemical Name or Material FAAH-IN-2
CAS 184475-71-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H11ClFN3O2
Quantity 5 g
Synonym O-Desmorpholinopropyl Gefitinib
Solubility Information DMSO : 20.83 mg/mL (65.15 mM; Need ultrasonic)
SMILES OC1=CC2=C(NC3=CC=C(F)C(Cl)=C3)N=CN=C2C=C1OC
Molecular Weight (g/mol) 319.72
Formula Weight 319.72
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.