Learn More
FABP3/H-FABP Antibody (FABP3/8343R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20519-20ug
 View more versions of this product
Product Code. 18638794
  • 286.00 € / 20µg
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FABP3/H-FABP
Monoclonal
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 μg/mL
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP
Human recombinant FABP3/H-FABP protein fragment (around aa1-127) (exact sequence is proprietary)
20 μg
Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism
2170
Store at 4&drg;C. Do not freeze.
IgG κ
Immunohistochemistry (Paraffin)
FABP3/8343R
Unconjugated
10mM PBS with 0.05% BSA
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified