missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FABP4 Recombinant Rabbit Monoclonal Antibody (1H4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560635
This item is not returnable.
View return policy
Description
FABP4 Recombinant Monoclonal Antibody for ELISA
FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.
Specifications
| FABP4 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15090 | |
| FABP4 | |
| A synthesized peptide derived from human FABP4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 1H4 | |
| Unconjugated | |
| FABP4 | |
| 3T3-L1 lipid-binding protein; 422/aP2; adipocyte fatty acid binding protein; Adipocyte lipid binding protein; adipocyte lipid-binding protein; adipocyte protein aP2; adipocyte-type fatty acid-binding protein; AFABP; A-FABP; ALBP; ALBP/Ap2; Ap2; epididymis secretory protein Li 104; FABP4; fatty acid binding protein 4; Fatty acid binding protein 4 adipocyte; fatty acid binding protein 4, adipocyte; fatty acid-binding protein 4; Fatty acid-binding protein, adipocyte; HEL S 104; HEL-S-104; Lbpl; myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2167 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction