missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP7/B-FABP Antibody (CL0236) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52889CL1
This item is not returnable.
View return policy
Description
FABP7/B-FABP Monoclonal antibody specifically detects FABP7/B-FABP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| FABP7/B-FABP | |
| Monoclonal | |
| CoraFluor 1 | |
| B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related | |
| This antibody was developed using a recombinant protein derived from O15540, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Cycle and Replication, Stem Cells | |
| 2173 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0236 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction