missing translation for 'onlineSavingsMsg'
Learn More

3M™ Face Seal

Product Code. 15719747 Shop All 3M Products
Change view
Click to view available options
Color: Black, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15719747 Color: Black, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15719747 Supplier 3M™ Supplier No. 7000044244
Quantity Price
1 40.17 € / Pack of 3
4 36.79 € / Pack of 3

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Face Seal for use with the 3M™ Visionair Supplied Air System.

  • Replacement face seal for use with the 3M™ Visionair Supplied Air System
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Approved
Color Black, Gray
For Use With (Equipment) Visionair Supplied Air System
Product Type Face Seal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.