missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor VII Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115207
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Factor VII.
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
Specifications
| Factor VII | |
| Polyclonal | |
| Unconjugated | |
| F7 | |
| AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14068, 2155, 260320 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08709, P70375, Q8K3U6 | |
| F7 | |
| A synthesized peptide derived from human F7(Accession P08709), corresponding to amino acid residues C187-L237. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?