Learn More
Factor VIII Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NB100-91761
 View more versions of this product
Product Code. 18727803
  • 565.00 € / 0.10mg
Description
Factor VIII Polyclonal specifically detects Factor VIII in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Factor VIII
F8
A synthetic peptide made to a C-terminal portion of human Factor VIII (between amino acids 2100-2250) [UniProt P00451]
0.1 mg
Mouse, Rat
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Rabbit
Affinity Purified