missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XII Antibody (OTI5A6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | Factor XII |
|---|---|
| Clone | OTI5A6 |
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306448
|
Novus Biologicals
NBP3-44901-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306682
|
Novus Biologicals
NBP3-44901-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Factor XII Monoclonal antibody specifically detects Factor XII in Human samples. It is validated for Western BlotSpecifications
| Factor XII | |
| Western Blot 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, F12, HAE3, HAEX, HAF, HAF beta-factor XIIa part 2, Hageman factor | |
| Human recombinant protein fragment corresponding to amino acids 73-372 of human F12(NP_000496) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI5A6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 2161 | |
| IgG2b | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title