missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIa Mouse, Clone: SPM180, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Factor XIIIa |
---|---|
Clone | SPM180 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959977
|
EnquireBio
2162-MSM2X-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949977
|
EnquireBio
2162-MSM2X-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Factor XIIIa | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
2162 | |
Recombinant fragment of human Factor XIIIa protein (aa46-181) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
83kDa |
SPM180 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_000120 | |
F13A1 | |
IgG2b κ | |
Protein A/G Purified | |
Cytoplasmic and Secreted | |
293T Cells. Placenta. |