missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99806-50ul
This item is not returnable.
View return policy
Description
Factor XIIIb Polyclonal antibody specifically detects Factor XIIIb in Human samples. It is validated for ELISA
Specifications
| Factor XIIIb | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| coagulation factor XIII B chain, coagulation factor XIII, B polypeptide, F13B, fibrin-stabilizing factor B subunit, FXIIIB, protein-glutamine gamma-glutamyltransferase B chain, TGase, transglutaminase B chain | |
| Produced in rabbits immunized with purified, recombinant Human Factor XIIIb (Accession#: P05160; Met1-Thr661) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 2165 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction