missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAIM3 Antibody (992338) [PE/Cy5.5], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ FAB9494PECY55
This item is not returnable.
View return policy
Description
FAIM3 Monoclonal antibody specifically detects FAIM3 in Human,Mouse,Rat samples. It is validated for Flow Cytometry
Specifications
| FAIM3 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, TOSOIgM Fc receptor | |
| Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession # O60667 | |
| 0.1 mL | |
| Apoptosis | |
| 9214 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 992338 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction