missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ FluoroBlok™96 Well HTS Insert Systems

Product Code. 10234152 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Unit Size
10234152 Flat Bottom (F-well) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10234152 Supplier Falcon™ Supplier No. 353928
  • ONLINE EXCLUSIVE PRICE
    208.00 €
    193.64 € / Pack of 5
    Save 14.36 €
    6% Off
Estimated Shipment: 09-04-2027
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify detection of migrating fluorescently labeled cells. Falcon™ FluoroBlok™96 Well HTS Insert Systems are designed with a unique light-tight PET membrane that blocks the transmission light within the range of 400 to 700nm.

96 square well, flat-bottom microplate
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Nonpyrogenic
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid With Lid
    Culture Area 0.08 cm2
    Array 8 x 12
    For Use With (Equipment) FluoroBlok™ 96 Well Multiwell HTS Insert System
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Untreated
    Well Shape Flat Bottom (F-well)
    Includes 96 square well, flat bottom plate
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.