missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM170A Antibody (OTI4C6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44989-100ul
This item is not returnable.
View return policy
Description
FAM170A Monoclonal antibody specifically detects FAM170A in Human samples. It is validated for Western Blot
Specifications
| FAM170A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 340069 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| OTI4C6 | |
| Western Blot 1:2000 | |
| family with sequence similarity 170, member A, Znf domain containing protein | |
| Full length human recombinant protein of human FAM170A (NP_877438) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction