missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM170A Antibody (OTI6E4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44995-30ul
This item is not returnable.
View return policy
Description
FAM170A Monoclonal antibody specifically detects FAM170A in Human samples. It is validated for Western Blot
Specifications
| FAM170A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 340069 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| OTI6E4 | |
| Western Blot 1:2000 | |
| family with sequence similarity 170, member A, Znf domain containing protein | |
| Full length human recombinant protein of human FAM170A(NP_877438) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto