missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM20B Monoclonal Antibody (1018569)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561952
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| FAM20B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75063 | |
| FAM20B | |
| Chinese hamster ovary cell line CHO-derived recombinant human FAM20B, Asn27-Leu409. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 1018569 | |
| Unconjugated | |
| FAM20B | |
| FAM20B; family with sequence similarity 20 member B; family with sequence similarity 20, member B; glycosaminoglycan xylosylkinase; gxk1; KIAA0475; Xylose kinase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9917 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction