missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM3A Polyclonal Antibody
GREENER_CHOICE

Product Code. 30534509
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534509 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534509 Supplier Invitrogen™ Supplier No. PA5148635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

A novel cytokine-like gene family using structure-based methods to search for novel four-helix-bundle cytokines in genomics databases. There are four genes in this family, FAM3A, FAM3B, FAM3C, and FAM3D, each encoding a protein (224-235 amino acids) with a hydrophobic leader sequence. Northern analysis indicates that FAM3B is highly expressed in pancreas, FAM3D in placenta, and FAM3A and FAM3C in almost all tissues. Immunohistochemistry showed that FAM3A is expressed prominently in the vascular endothelium, particularly capillaries. Fam3a staining in capillary endothelium and endocardium of heart, as well as vascular endothelial staining in many other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM3A
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene FAM3A
Gene Accession No. P98173
Gene Alias 1810037C20Rik; 2.19; 2-19; Cytokine-like protein 2-19; DLD; DXS560S; FAM3 metabolism regulating signaling molecule A; FAM3A; family 3, member A protein; family with sequence similarity 3 member A; family with sequence similarity 3, member A; hypothetical protein MGC73350; protein FAM3A; RGD1562076; XAP-7; zgc:73350
Gene Symbols FAM3A
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human FAM3A, Phe34-Ser230.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.