missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM3B Monoclonal Antibody (365505)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561214
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
FAM3B (family with sequence similarity 3, member B), also known as ORF9, C21orf11 or PANDER, is a 235 amino acid secreted protein that localizes to discrete vesicular and perinuclear structures. Expressed at high levels in pancreas and at lower levels in kidney, colon, testis, prostate and small intestine, FAM3B functions as an islet-specific cytokine that promotes apoptosis and may inhibit Insulin secretion from beta-cells, possibly playing a role in overall pancreatic activity. Human FAM3B shares 78% sequence identity with its mouse counterpart, suggesting a conserved role between species. Three isoforms of FAM3B, designated A, B and C, exist due to alternative splicing events.
Specifications
| FAM3B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P58499 | |
| FAM3B | |
| Mouse myeloma cell line NS0-derived recombinant human FAM3B isoform B, Glu30-Ser235. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 365505 | |
| Unconjugated | |
| FAM3B | |
| 21-Feb; 2-21; 9030624C24Rik; C21orf11; C21orf76; cytokine-like protein 2-21; D16Jhu19e; D21M16SJHU19e; FAM3 metabolism regulating signaling molecule B; Fam3b; family with sequence similarity 3 member B; family with sequence similarity 3, member B; ORF9; pancreatic derived factor; pancreatic-derived factor; PANDER; PRED44; Protein FAM3B; RGD1311662; UNQ320/PRO365 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 54097 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction