missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM50B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 589.00 €
Specifications
| Antigen | FAM50B |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681649
|
Novus Biologicals
NBP2-49529-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639046
|
Novus Biologicals
NBP2-49529 |
0.1 mL |
589.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM50B Polyclonal antibody specifically detects FAM50B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FAM50B | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| family with sequence similarity 50, member B, Protein XAP-5-like, X5LD6S2654E | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QEALVRERERQLAKRQHLEEQRLQQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 26240 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title