missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM83C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37925-25ul
This item is not returnable.
View return policy
Description
FAM83C Polyclonal specifically detects FAM83C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FAM83C | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9BQN1 | |
| FAM83C | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VGDPDSGVTPNSGPLRPGEQAPEDRRLSPSQADSQLDLLSRALGTGGAPELGSLRPGDRALEDRRLSLNQSRGQSDLLMQYPKAQGSRVPLETNSSARP | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C20orf128, Chromosome 20 Open Reading Frame 128, dJ614O4.7, EC 1.6.5, EC 6.3.5.3, Family With Sequence Similarity 83, Member C, Protein FAM83C | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 128876 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction