missing translation for 'onlineSavingsMsg'
Learn More

Famotidine, MedChemExpress

Product Code. 16480898
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480898 5 g 5g
16402159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480898 Supplier Medchem Express Supplier No. HYB0377
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Famotidine (MK-208) is a competitive histamine H2-receptor antagonist. Its main pharmacodynamic effect is the inhibition of gastric secretion.

Famotidine (MK-208) is a competitive histamine H2-receptor antagonist. Its main pharmacodynamic effect is the inhibition of gastric secretion.

Specifications

Chemical Name or Material Famotidine
Physical Form Solid
CAS 76824-35-6
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H15N7O2S3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym MK-208
Solubility Information DMSO : ≥ 100 mg/mL (296.34 mM) ∣H2O : 0.1 mg/mL (0.30 mM; Need ultrasonic)
SMILES N=C(NS(=O)(N)=O)CCSCC1=CSC(NC(N)=N)=N1
Molecular Weight (g/mol) 337.45
Formula Weight 337.45
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.