missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAS Monoclonal Antibody (DX2), Pacific Blue™, Invitrogen™

Mouse Monoclonal Antibody
483.00 €
Specifications
| Antigen | FAS |
|---|---|
| Clone | DX2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Pacific Blue |
Description
FAS (Fas, CD95, APO-1) is a 46 kDa transmembrane glycoprotein that functions as a cell death receptor of the TNFR (tumor necrosis factor receptor) superfamily. FAS also exists in a soluble form that weight approximately 26kDa. Stimulation of FAS results in aggregation of its intracellular FAS-associated death domains (FADD), formation of the death-inducing signaling complex (DISC) and activation of caspases. The auto-proteolytic processing of the caspases in the complex trigger a downstream cascade that leads to apoptosis. FAS has been shown to activate NF-kappaB, MAPK3/ERK1, MAPK8/JNK, and is involved in transducing the proliferating signals in normal diploid fibroblast and T cells. At least eight alternatively spliced transcript variants encoding seven distinct isoforms have been described. The isoforms lacking the transmembrane domain may negatively regulate the apoptosis mediated by the full-length isoform. The Fas/Fas ligand system has been shown to play a role in a number of human diseases such as AIDS, hepatitis and cancer. It is also assumed that induction of apoptosis through FAS ligand pathway is predominantly involved in anti-viral immune responses.Specifications
| FAS | |
| Flow Cytometry | |
| Pacific Blue | |
| Mouse | |
| Human | |
| 355 | |
| Human CD95. | |
| Primary | |
| Antibody |
| DX2 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P25445 | |
| FAS | |
| IgG1 | |
| 4°C | |
| FAS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title