missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastCellular™ Senescence Detection Kit, SPiDER-ΒGal

Product Code. 16359855 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Application):
Cellular Senescence Detection
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
16359855 Cellular Senescence Detection 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16359855 Supplier MP Biomedicals™ Supplier No. 092690301
  • ONLINE EXCLUSIVE PRICE
    368.00 €
    348.60 € / 1 sets
    Save 19.40 €
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect senescent cells with much higher sensitivity than X-gal.

Cellular senescence is a phenomenon by which normal cells cease to divide and enter a state of irreversible growth arrest. It is one of the most fundamental aspects of cellular behavior and generally believed to be associated with tumor suppressive mechanisms and an underlying cause of aging. Senescence-associated Β-galactosidase (SA-Β-gal), which is overexpressed in senescent cells, has been widely used as a marker of cellular senescence. Although X-gal is a well-known reagent used to detect SA-Β-gal, X-gal has the following disadvantages: 1) requirement of fixed cells due to poor cell-permeability, 2) low quantitative capability because of the difficulty in visually differentiating stained vs. non-stained cells 3) requirement for long stain times.

To conquer the disadvantages inherent in using X-gal, we utilize a new reagent and method for the detection of cellular senescence. SPiDER-ΒGal offers increased sensitivity in detecting SA-Β-gal due to increased cell permeability and retention inside cells. Compared to the X-gal method, the FastCellular Senescence Detection Kit can be used for both living and fixed cells, because we provide Bafilomycin A1 to specifically inhibit endogenous Β-galactosidase activity in living cells and thus increase the detection specificity for living cells. For fixed cells, SA-Β-gal is detectable by using SPiDER-ΒGal and Mcllvaine buffer (pH 6.0, please refer to the protocol for fixed cell assays). Since SPiDER-ΒGal emits strong and stable fluorescence after the reaction with SA-Β-gal, it can be applied to quantitative analysis by flow cytometry. In addition, the staining time is only 30 mins. Kit includes a 6-well plate with 1 unit of SPiDER-ΒGal and 1 unit of Bafilomycin A1

Benefits of using FastCellular Senescence Detection Kit:

  • Higher sensitivity due to a new fluorogenic detection probe
  • Applicable for living cells and fixed tissues
  • Ability to quantify SA-Β-gal in cellular senescence assay
  • Short staining time (30 min)

Specifications

Content And Storage Store at 0 to 5°C with protection from light
For Use With (Application) Cellular Senescence Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.