missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fastidious Anaerobe Agar (F.A.A.) with Horse Blood

Product Code. 12957138
Change view
Click to view available options
19mL; 1 x 10 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12957138 19mL; 1 x 10 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12957138 Supplier Thermo Scientific™ Supplier No. PB0225A
  • ONLINE EXCLUSIVE PRICE
    28.65 €
    23.58 € / Pack of 10
    Save 5.07 €
    17% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow most clinically significant fastidious anaerobes with this primary isolation medium.

Grow most clinically significant anaerobes with Thermo Scientific™ Fastidious Anaerobe Agar (F.A.A.) with Horse Blood, a primary isolation medium for the recovery of fastidious anaerobic bacteria.

Fastidious Anaerobe Agar (F.A.A.) with Horse Blood is a medium for the growth of fastidious organisms. The peptones included have been chosen for maximum growth stimulation. Starch and sodium bicarbonate act as detoxification agents while haemin encourages pigment production in Porphyromonas melaninogenicus. Specific growth promoting agents are Cysteine for Fusobacterium necrophorum, Propionibacterium acne and Bacteroides fragilis, arginine for Eubacterium spp., and soluble pyrophosphate for Porphyromonas gingivalis and Porphyromonas asaccharolyticus. Pyruvate helps neutralize hydrogen peroxide and is also utilized by Veillonella spp. as an energy source. Vitamin K and sodium succinate provide essential growth factors for some anaerobes, as does the 0.1% glucose. The low level of glucose prevents the production of high levels of acids and alcohols which would inhibit colonial development.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 10°C
Description Fastidious Anaerobe Agar (F.A.A.) with Horse Blood
Format 90mm Monoplate
Product Type Agar Plate
Fill Volume 19 mL
Packaging Type 1 x 10 plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.