Learn More
Thermo Scientific™ Fastidious Anaerobe Agar with Neomycin
Description
Selectively isolate most clinically significant anaerobes with Thermo Scientific™ Fastidious Anaerobe Agar with Neomycin. Anaerobes are widely distributed in nature and are the predominant components of bacterial flora of normal human skin and mucous membranes. Fastidious Anaerobe Agar with Neomycin is a primary isolation medium which includes specific peptones for the recovery of fastidious anaerobic bacteria.
Fastidious Anaerobe Agar with Neomycin includes a number of peptones chosen for maximum growth stimulation of fastidious organisms. Starch and sodium bicarbonate act as detoxification agents, while haemin encourages pigment production in Porphyromonas melaninogenicus. Specific growth-promoting agents are cysteine for Fusobacterium necrophorum, Propionibacterium acne and Bacteriodes fragilis; arginine for Eubacterium spp.; and soluble pyrophosphate for Porphyromonas gingivalis and Porphyromonas asaccharolyticus. Pyruvate helps neutralize hydrogen peroxide and is also utilized by Veillionella spp. as an energy source. Vitamin K and sodium succinate provide essential growth factors for some anaerobes as does the 0.1% glucose. The low level of glucose prevents the production of high levels of acids and alcohols which would inhibit colonial development.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 10°C |
| Description | F.A.A. with Neomycin |
| Format | 90mm Monoplate |
| Product Type | Agar Plate |
| Fill Volume | 19 mL |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.