missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastMitophagy™ Detection Kit

Product Code. 16301296 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 μg, 30μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16301296 5 μg, 30μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16301296 Supplier MP Biomedicals™ Supplier No. 092690201
  • ONLINE EXCLUSIVE PRICE
    794.00 €
    752.85 € / 1 sets
    Save 41.15 €
    5% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for mitophagy detection in mammalian cells.

The FastMitophagy Detection Kit is specifically designed for mitophagy detection in mammalian cells. Mitophagy is the selective degradation of defective mitochondria by autophagy following DNA damage or oxidative stress. It serves as a specific elimination system by sequestering dysfunctional mitochondria into autophagosomes, fusing the phagosome to lysosomes, and degrading them by digestion. Since its first observation in 1962, mitophagy has attracted intense research for cell homeostasis and development in mammalian cells. Recent research efforts are directed towards further elucidating the role of induced autophagy as a cell survival response to stress, microbial infection, and disease (e.g., cancer, Alzheimer's disease, and Parkinson's disease).
This kit is composed of a mitophagy dye, a reagent for detection of mitophagy, and lysosome dye (Lyso dye). Mitophagy dye initially accumulates in intact mitochondria and becomes immobilized via chemical bonding, resulting in a very weak fluorescence due to the influence of the surrounding environment. When mitophagy is induced, the damaged mitochondria fuses to lysosome and the Mitophagy dye emits a high fluorescence. Lyso dye is used to confirm the fusion of Mitophagy dye - labeled mitochondria and lysosome.
Each kit contains 5 μg of Mitophagy Dye and 30 μg Lyso Dye which is enough for 5 x 96 well plates or 25 assays when using a 35 mm.

Benefits of using FastMitophagy Detection Kit:

  • Simple procedure: just add the Mitophagy dye - no need for transfection
  • More sensitive than other autophagy markers

Specifications

Content And Storage Store at 0 to 5°C with protection from light
For Use With (Application) Mitophagy Detection Kit
Includes Each kit contains 5 µg of Mitophagy Dye and 30 µg Lyso Dye which is enough for 5 x 96 well plates
Quantity 5 μg, 30μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.