missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP4/SLC27A4 Antibody (S01-3C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19773-100ul
This item is not returnable.
View return policy
Description
FATP4/SLC27A4 Monoclonal antibody specifically detects FATP4/SLC27A4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| FATP4/SLC27A4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10999 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| S01-3C7 | |
| Unconjugated | |
| ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4 | |
| Recombinant protein of human FATP4/SLC27A4 (Uniprot # Q6P1M0) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction