missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBP1 Recombinant Rabbit Monoclonal Antibody (K01_3J78)

Product Code. 30830364
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830364 50 μL
30828553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830364 Supplier Invitrogen™ Supplier No. MA565769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

FBP1 Recombinant Monoclonal Antibody for Western Blot

Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBP1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3J78
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene FBP1
Gene Accession No. P09467, P19112, Q9QXD6
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; Fbp; FBP 1; FBP1; Fbp2; Fbp-2; Fbp3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fdp; fructose bisphosphatase 1; fructose-1; fructose-1,6- biphosphatase 1; fructose-1,6-bisphosphatase; fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; fructose-bisphosphatase 1; growth-inhibiting protein 17; liver FBPase
Gene Symbols FBP1
Host Species Rabbit
Immunogen Peptide.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14121, 2203, 24362
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.