Learn More
FBXL19 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23624-1-AP-20UL
This item is not returnable.
View return policy
Description
The FBXL19 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FBXL19. This antibody recognizes human antigen. The FBXL19 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FBXL19 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| F box/LRR repeat protein 19, Fbl19, FBXL19, JHDM1C | |
| 20 μL | |
| 54620 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |