missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO15 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149123
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
F-box protein 15 (FBXO15) is a gene located on chromosome 20p11.1 that encodes a member of the F-box protein family, known for their role in ubiquitin-mediated proteolysis. Proteins in this family contain an F-box domain, enabling them to form part of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex, which targets specific proteins for ubiquitination and subsequent degradation. FBXO15 is involved in regulating various cellular processes by controlling the turnover of proteins related to cell cycle progression, signal transduction, and development. Its function is crucial for maintaining cellular homeostasis, where dysregulation of FBXO15 can impact protein degradation pathways, potentially leading to disorders such as cancer due to uncontrolled cell proliferation. Current research is focused on understanding the specific targets and functions of FBXO15 within the ubiquitin proteasome system and its potential as a therapeutic target in diseases characterized by aberrant proteolytic activity.
Specifications
| FBXO15 | |
| Polyclonal | |
| Unconjugated | |
| FBXO15 | |
| AU019763; ecat3; F-box only protein 15; F-box protein 15; FBX15; FBXO15; MGC39671 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 201456 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8NCQ5 | |
| FBXO15 | |
| E. coli-derived recombinant human F-box protein 15/FBXO15, aa 298-434. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction