missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (439-9B), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43369APCCY7
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Bovine samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ITGB4 | |
| The immunogen for this antibody was CD104. | |
| 0.1 mL | |
| Primary | |
| Human, Bovine | |
| IgG2b |
| Flow Cytometry | |
| 439-9B | |
| Flow Cytometry | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Rat | |
| Protein A or G purified | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3691.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction