missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI alpha Antibody (Mar-1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09005-0.2mg
This item is not returnable.
View return policy
Description
Fc epsilon RI alpha Monoclonal antibody specifically detects Fc epsilon RI alpha in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Specifications
| Fc epsilon RI alpha | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| 2024-03-01 00:00:00 | |
| Unconjugated | |
| FCE1A;fcepsilonri;FCER1A;FcERI;Fcr-5 | |
| FceR1 alpha | |
| 0.2 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction