Learn More
TGF-beta Antibody (TGFB/510), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08739PCP
Product Code. 18684587
  • 736.00 € / 0.10mL
Description
TGF-beta Monoclonal antibody specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGF-beta
Monoclonal
PerCP
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Full length recombinant human TGF-beta protein
0.1 mL
Cytokine Research, Extracellular Matrix
7040
Store at 4°C in the dark.
IgG1 κ
ELISA, Functional Assay
TGFB/510
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat, Bovine, Canine
Purified