missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIII (CD16) Antibody (ICO-116), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
584.00 € - 631.00 €
Specifications
| Antigen | Fc gamma RIII (CD16) |
|---|---|
| Clone | ICO-116 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687857
|
Novus Biologicals
NBP2-47955B-100ul |
100 μg |
631.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644576
|
Novus Biologicals
NBP2-47955B |
0.1 mL |
584.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIII (CD16) Monoclonal antibody specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| Fc gamma RIII (CD16) | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| PBS | |
| 2214 | |
| IgG1 κ | |
| Protein A or G purified |
| ICO-116 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Human Fc gamma RIII (CD16) protein (Uniprot: P08637) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title