missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCAR/CD89 Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89482-100UL
This item is not returnable.
View return policy
Description
FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| FCAR/CD89 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2204 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 2 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
| Recombinant Human FCAR/CD89 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction