missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCGR1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526205
This item is not returnable.
View return policy
Description
FCGR1B Polyclonal Antibody for Western Blot, IHC (P), Flow
FCGR1B may bind to the Fc region of immunoglobulins gamma with a low affinity compared to FCGR1A. This protein may function in the humoral immune response.
Specifications
| FCGR1B | |
| Polyclonal | |
| Unconjugated | |
| FCGR1B | |
| CD64b; Fc fragment of IgG receptor Ib; Fc fragment of IgG, high affinity Ib, receptor (CD64); Fc gamma receptor Ib; Fc-gamma receptor I B2; fc-gamma RIB; FCGR1B; FcRIB; hFcgammaRIB; high affinity immunoglobulin gamma Fc receptor IB; IGFRB; IgG Fc receptor IB | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 2210 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q92637 | |
| FCGR1B | |
| KLH conjugated synthetic peptide between 248-276 amino acids from the C-terminal region of human FCGR1B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction