Learn More
FCHO2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558060
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KAAVKSKKAT DTYKLYVEKY ALAKADFEQK MTETAQKFQD IEETHLIHIK EIIGSLSNAI KEIHLQIG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
FCHO2 (FCH domain only 2) is a ubiquitously expressed member of the FCFBS superfamily characterized by FES-CIP4 homology (FCH) domain, an FBH domain, and an SH3 domain. Both FCHO2 and the related protein FCHO1 are mediators of clathrin-mediated endocytosis, acting to sculpt the initial bud site and recruit the clathrin machinery for clathrin-coated vesicle formation. FCHO2 binds to Eps15, an important adaptor protein in clathrin-mediated endocytosis.Specifications
FCHO2 | |
Polyclonal | |
Unconjugated | |
FCHO2 | |
5832424M12Rik; AA387320; F-BAR domain only protein 2; FCH domain only 2; FCH domain only protein 2; FCHO2; RGD1565396 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115548 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0JRZ9 | |
FCHO2 | |
Recombinant protein corresponding to Human FCHO2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only