missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26949-1-AP-20UL
This item is not returnable.
View return policy
Description
Receptors for the Fc fragment of IgG, or FCGRs, are cell surface glycoproteins of the Ig superfamily (IgSF). These receptors mediate phagocytosis of IgG-coated pathogens and promote activation of effector cells, leading to inflammatory responses and antibody-mediated cellular cytotoxicity. All FCGR genes map to human chromosome 1. Additional genes in this region, including FREB, encode FCGR homologs that are selectively expressed in B cells and may be implicated in B-cell development and lymphomagenesis.Specifications
| FCRLA | |
| Polyclonal | |
| Unconjugated | |
| FCRLA | |
| Fc receptor like A, Fc receptor like protein, Fc receptor related protein X, FCRL, FCRL1, FCRLA, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLM1, FCRLX, FCRX, FREB, RP11 474I16.5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84824 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7L513 | |
| FCRLA | |
| FCRLA Fusion Protein Ag25331 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction