missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FCRN/FCGRT Antibody (rozanolixizumab) - Humanized, IgG4SP, Novus Biologicals™

Human Monoclonal Antibody

213.00 € - 920.00 €

Specifications

Antigen FCRN/FCGRT
Clone rozanolixizumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30046014
View Documents
Novus Biologicals™
NBP3-28355-100ug
100 μg
326.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045086
View Documents
Novus Biologicals™
NBP3-28355-50ug
50 μg
213.00 €
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30047320
View Documents
Novus Biologicals™
NBP3-28355-1mg
1 mg
920.00 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

FCRN/FCGRT
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Immunology
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
FcRn (FCGRT & B2M)
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
rozanolixizumab
Monoclonal
Purified
RUO
Human
P55899
2217
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.