Learn More
FDC Antibody (CNA.42), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-43118-0.025MG
 View more versions of this product
Product Code. 18265235
  • 325.00 € / 25µg
Description
FDC Monoclonal specifically detects FDC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDC
Monoclonal
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC
Mouse
Affinity Purified
RUO
1218
Store at 4C. Do not freeze.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
CNA.42
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CMD1B
The immunogen for this antibody was follicular dendritic cells.
0.025 mg
Primary
Human
IgM